1-Iodohexadecane ≥98% stabilised

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


Stabilisatie: Stabilised with copper

A12977.14 A12977.22 A12977.30
A12977.14EA 42.4 EUR
A12977.14 A12977.22 A12977.30
1-Iodohexadecane ≥98% stabilised
1-Iodohexadecane

Stabilized with copper

Formule: C₁₆H₃₃I
Kookpunt: 206…207 °C (10 mmHg)
Smeltpunt: 21…23 °C
Dichtheid: 1,121
Vlampunt: >110 °C (230 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001089
CAS nummer: 544-77-4
EINECS: 208-879-4
UN: 3082
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR