N,N-Dimethyldodecylamine 95% pract.
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
408290010 408292500
ACRO408290010EA 667 EUR
ACRO408290010 ACRO408292500
N,N-Dimethyldodecylamine 95% pract.
N,N-Dimethyldodecylamine
Formule: C₁₄H₃₁N
Kookpunt: 265 °C (1013 hPa)
Smeltpunt: –20 °C
Dichtheid: 0,787 g/cm³
Vlampunt: 118 °C
MDL Nummer: MFCD00008970
CAS nummer: 112-18-5
EINECS: 203-943-8
UN: 2735
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear to slightly hazy Liquid
Infrared spectrum Conforms
Refractive index 1.4360 to 1.4390 (20°C, 589 nm)
GC ≥94 % (Dodecyl)
Color scale ≤30 APHA
Titration ≥94 % (Tertiary amine)