Bis(2-hydroxyethyl)amino tris(hydroxymethyl)methane (BIS-TRIS) 99+%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
168871000 168870025 168875000
ACRO168871000EA 154 EUR
ACRO168871000 ACRO168870025 ACRO168875000
Bis(2-hydroxyethyl)amino tris(hydroxymethyl)methane (BIS-TRIS) 99+%
Bis(2-hydroxyethyl)amino tris(hydroxymethyl)methane (BIS-TRIS)
Formule: C₈H₁₉NO₅
Smeltpunt: 103…104 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002853
CAS nummer: 6976-37-0
EINECS: 230-237-7
ADR: 8,III
Order Now
Specification Test Results
Appearance White Powder
Infrared spectrum Conforms
Water ≤1.0 % (K.F.)
Titration with HClO4 ≥99.0 %