Bis(2-hydroxyethyl)amino tris(hydroxymethyl)methane (BIS-TRIS), Millipore®
Leverancier: Merck Millipore Calbiochem
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: 2,2-Bis(hydroxymethyl)-2,2',2''-nitrilotriethanol, Bis(2-hydroxyethyl)amino-tris(hydroxymethyl)methane (BIS-TRIS), 2-(Bis(2-hydroxyethyl)amino)-2-(hydroxymethyl)propane-1,3-diol, 2-[Bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
391335-1 391335-100
391335-1EA 954 EUR
391335-1 391335-100
Bis(2-hydroxyethyl)amino tris(hydroxymethyl)methane (BIS-TRIS), Millipore®
Bis(2-hydroxyethyl)amino tris(hydroxymethyl)methane (BIS-TRIS)
Formule: C₈H₁₉NO₅
Smeltpunt: 102…103 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002853
CAS nummer: 6976-37-0
EINECS: 230-237-7
Order Now
Specificaties
Environmentally Preferable