N,N,N',N'-Tetraethylethyleendiamine 99+%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
155590100 155591000 155595000
ACRO155590100EA 48.7 EUR
ACRO155590100 ACRO155591000 ACRO155595000
N,N,N',N'-Tetraethylethyleendiamine 99+%
N,N,N',N'-Tetraethylethyleendiamine
Formule: C₁₀H₂₄N₂
Kookpunt: 189…192 °C (760 mmHg)
Dichtheid: 0,808 g/cm³
Vlampunt: 58 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009055
CAS nummer: 150-77-6
EINECS: 205-770-3
UN: 2734
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4333 to 1.4353 (20°C, 589 nm)
GC ≥99.0 %