Hexamethyleen-1,6-diisocyanaat ≥98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Hexamethylene diisocyanate
Packaged under argon.
43803.30
43803.18
43803.30EA
113
EUR
43803.30
43803.18
Hexamethyleen-1,6-diisocyanaat ≥98%
Hexamethyleen-1,6-diisocyanaat
Purity: 98+%
CAS No.: 822-06-0
Molecular Formula: OCN(CH2)6NCO
Formula Weight: 168.19
Flash Point: 130°(266°F)
Physical Form: Liquid, Packaged under argon
Density: 1.05
Refractive Index: 1.4520
Sensitivity: Moisture Sensitive
Odour: Pungent
Melting Point: -67°
Boiling Point: 255°
Beilstein Reference: 956709
UN No.: 2281
MDL No.: MFCD00002047
Formule:
C₈H₁₂N₂O₂ Kookpunt: 255 °C Smeltpunt: –67 °C Dichtheid: 1,05 Vlampunt: 130 °C (266 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00002047 CAS nummer: 822-06-0 EINECS: 212-485-8 UN: 2281 ADR: 6.1,II |