1-Pentyl isothiocyanate
Leverancier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TL04631.1
TL04631EA
TL04631.50
MAYBTL04631.1EA
0
EUR
MAYBTL04631.1
MAYBTL04631EA
MAYBTL04631.50
1-Pentyl isothiocyanate
1-Pentyl isothiocyanate
Formule:
C₆H₁₁NS Kookpunt: 191 °C (1013 hPa) Dichtheid: 0,93 g/cm³ (20 °C) |
MDL Nummer:
MFCD00014444 CAS nummer: 629-12-9 EINECS: 211-075-6 UN: 2922 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...