1,4-Butane diisothiocyanate ≥98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
L09234.03
L09234.06
L09234.03EA
63.9
EUR
L09234.03
L09234.06
1,4-Butane diisothiocyanate ≥98%
1,4-Butane diisothiocyanate
Purity: 98%
CAS No.: 4430-51-7
Molecular Formula: SCN(CH2)4NCS
Formula Weight: 172.27
Odour: Irritating
Melting Point: 30-33°
Boiling Point: 170°/11mm
Beilstein Reference: 1757138
UN No.: 2923
MDL No.: MFCD00041136
Formule:
C₆H₈N₂S₂ Kookpunt: 170 °C (11 mmHg) Smeltpunt: 30…33 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00041136 CAS nummer: 4430-51-7 UN: 2923 ADR: 8,III |