2-(2-Aminoethoxy)ethanol for synthesis, Sigma-Aldrich®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
M-Clarity™ quality level = MQ200
8.18431.0100 8.18431.0500
8.18431.0100EA 43.3 EUR
8.18431.0100 8.18431.0500
2-(2-Aminoethoxy)ethanol for synthesis, Sigma-Aldrich®
2-(2-Aminoethoxy)ethanol
Formule: C₄H₁₁NO₂
MW: 105.14 g/mol
Kookpunt: 220
Smeltpunt: –11 °C
Dichtheid: 1.0966 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008181
CAS nummer: 929-06-6
EINECS: 213-195-4
UN: 3055
ADR: 8 III
Order Now
Specification Test Results
Solubility (20 °C) soluble
Melting Point -11 °C
Molar Mass 105.14 g/mol
Boiling Point 125 - 131 °C (30 hPa)
Vapor Pressure  - 0.1 hPa (20 °C)
Flash Point 127 °C
Explosion Limit 2.0 - 15.5 %(V)
Density 1.06 g/cm3 (20 °C)
pH 10.2 (10 g/l, H2O, 20 °C)
Ignition Point 370 °C