Tris-(hydroxymethyl)-aminomethaan (TRIS) ACS, Reag. Ph. Eur. analytical reagent, buffer substance, Millipore®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ400
1.08382.0100 1.08382.0500 1.08382.1000 1.08382.2500 1.08382.9025
1.08382.0100EA 59.5 EUR
1.08382.0100 1.08382.0500 1.08382.1000 1.08382.2500 1.08382.9025
Tris-(hydroxymethyl)-aminomethaan (TRIS) ACS, Reag. Ph. Eur. analytical reagent, buffer substance, Millipore®
Tris-(hydroxymethyl)-aminomethaan (TRIS)
Formule: NH₂C(CH₂OH)₃
Kookpunt: 357 °C (1013 hPa)
Smeltpunt: 169…172 °C
Dichtheid: 1,328 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004679
CAS nummer: 77-86-1
EINECS: 201-064-4
Merck Index: 13,09842
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11a03d1ab6b2c11b9a2c83f0a73fc4264cec0110&vid=5ce890cdf298a59a7900a07fee1c857a5e5e61e0&prid=web&clid=SAPDAM