Tris-(hydroxymethyl)-aminomethaan (TRIS), Millipore®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: 2-Amino-2-(hydroxymethyl)propane-1,3-diol(tris), Tromethamine, Tris(hydroxymethyl)methylamine, 2-Amino-2-(hydroxymethyl)propane-1,3-diol, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, Trizma(R) base, TRIS base, THAM
M-Clarity™ quality level = MQ100
1.08387.0500 1.08387.2500
1.08387.0500EA 68.4 EUR
1.08387.0500 1.08387.2500
Tris-(hydroxymethyl)-aminomethaan (TRIS), Millipore®
Tris-(hydroxymethyl)-aminomethaan (TRIS)
Formule: NH₂C(CH₂OH)₃
MW: 121.14 g/mol
Kookpunt: 357
Smeltpunt: 169…172 °C
Dichtheid: 1.328 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004679
CAS nummer: 77-86-1
EINECS: 201-064-4
Merck Index: 13,09842
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11a03d1ab6b2c11b9a2c83f0a73fc4264cec0110&vid=5ce890cdf298a59a7900a07fee1c857a5e5e61e0&prid=web&clid=SAPDAM