Dioctylamine 96%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

A18017.14 A18017.22
A18017.14EA 35.9 EUR
A18017.14 A18017.22
Dioctylamine 96%
Dioctylamine

Purity: 98% CAS No.: 1120-48-5 Molecular Formula: C16H35N Formula Weight: 241.46 Flash Point: 142°(287°F) Refractive Index: 1.4432 Odour: Amine-like Melting Point: 13-15° Boiling Point: 297-298° Beilstein Reference: 1748376 UN No.: 2735 MDL No.: MFCD00009557

Formule: C₁₆H₃₅N
Kookpunt: 297…298 °C
Smeltpunt: 13…15 °C
Dichtheid: 0,799
Vlampunt: 142 °C (287 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009557
CAS nummer: 1120-48-5
EINECS: 214-311-6
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR