2-Ethylaminoethanol 98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
N-Ethyl-2-aminoethanol, N-Ethyl-2-ethanolamin, 2-(Ethylamino)ethan-1-ol
A15744.AE
A15744.AP
A15744.0F
A15744.AEEA
25.9
EUR
A15744.AE
A15744.AP
A15744.0F
2-Ethylaminoethanol 98%
2-Ethylaminoethanol
Purity: 98%
CAS No.: 110-73-6
Molecular Formula: C4H11NO
Formula Weight: 89.14
Flash Point: 71°(160°F)
Refractive Index: 1.441
Melting Point: -9°
Boiling Point: 168-170°
Beilstein Reference: 635671
UN No.: 2922
MDL No.: MFCD00002841
Formule:
C₄H₁₁NO Kookpunt: 168…170 °C Smeltpunt: –9 °C Dichtheid: 0,914 Vlampunt: 71 °C (160 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00002841 CAS nummer: 110-73-6 EINECS: 203-797-5 UN: 2922 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...