2-(Cyclohexylamino)ethanesulphonic acid (CHES) 98+% for biochemistry
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: N-Cyclohexyltaurine, CHES
208181000 208180250 208185000
ACRO208181000EA 105 EUR
ACRO208181000 ACRO208180250 ACRO208185000
2-(Cyclohexylamino)ethanesulphonic acid (CHES) 98+% for biochemistry
2-(Cyclohexylamino)ethanesulphonic acid (CHES)
Formule: C₈H₁₇NO₃S
MW: 207.29 g/mol
Smeltpunt: 300 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003835
CAS nummer: 103-47-9
EINECS: 203-115-6
Merck Index: 13,02063
Order Now
Specification Test Results
Appearance White Crystalline powder
Infrared spectrum Conforms
Water ≤1 % (K.F.)
UV at 280 nm A: ≤0.04
pH 5 to 6 (1 % in water)
Titration with NaOH ≥98.0 %