Ethyleenglycolmonomethylether (2-Methoxyethanol) ≥99% for spectrophotometry
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Ethylene glycol monomethyl ether
32444.K2
32444.M1
32444.K2EA
80.8
EUR
32444.K2
32444.M1
Ethyleenglycolmonomethylether (2-Methoxyethanol) ≥99% for spectrophotometry
Ethyleenglycolmonomethylether (2-Methoxyethanol)
Purity: 99% min
CAS No.: 109-86-4
Molecular Formula: C3H8O2
Formula Weight: 76.10
Flash Point: 46°(115°F)
Physical Form: Liquid
Density: 0.965
Refractive Index: 1.4025
Appearance: Colourless
Odour: Characteristic
pH: 4-7
Melting Point: -85°
Boiling Point: 124-125°
Beilstein Reference: 1731074
Merck Reference: 14,6038
UN No.: 1188
MDL No.: MFCD00002867
Formule:
C₃H₈O₂ Kookpunt: 124…125 °C Smeltpunt: –85 °C Dichtheid: 0,965 Vlampunt: 46 °C (115 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00002867 CAS nummer: 109-86-4 EINECS: 203-713-7 UN: 1188 ADR: 3,III Merck Index: 14,6038 |