VWR®, overall, met capuchon en ritssluiting
Leverancier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
113-9038EA 401 EUR
113-9038 113-9039 113-9040 113-9041 113-9042 113-9043 113-9044 113-9045 113-9046 113-9047
VWR®, overall, met capuchon en ritssluiting
Wegwerpkleding
These disposable overalls, with hood, are made of breathable and durable non woven SMS fabric.
  • Hood
  • Zip closure (one direction)
  • Elasticated wrists
  • Free from natural rubber latex
Weight: 50 g/m²
Verpakking: 1 piece per bag, 50 bags per case.