Azijnzuuranhydride 97+% ACS
Leverancier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Purity: 97+%
CAS No.: 108-24-7
Molecular Formula: C4H6O3
Formula Weight: 102.09
Flash Point: 49°(120°F)
Physical Form: Liquid
Density: 1.082
Refractive Index: 1.3900
Sensitivity: Moisture Sensitive
Appearance: Colourless
Odour: Pungent
Melting Point: -73°
Boiling Point: 138-140°
Beilstein Reference: 385737
Merck Reference: 14,56
UN No.: 1715
MDL No.: MFCD00008705
Formule:
(CH₃CO)₂O Kookpunt: 138…140 °C Smeltpunt: –73 °C Dichtheid: 1,082 Vlampunt: 49 °C (120 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00008705 CAS nummer: 108-24-7 EINECS: 203-564-8 UN: 1715 ADR: 8,II Bereik: 01-2119486470-36 Merck Index: 14,56 |