Dithiothreitol (DTT, Cleland's reagent), Millipore®
Leverancier: Merck Millipore Calbiochem
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: cis-2,3-Dihydroxy-1,4-butanedithiol, (R*,R*)-1,4-Dimercapto-2,3-butandiol, DTT, DL-1,4-Dithiothreitol
233156-1 233156-10
233156-1EA 84.1 EUR
233156-1 233156-10
Dithiothreitol (DTT, Cleland's reagent), Millipore®
Dithiothreitol (DTT, Cleland's reagent)
Formule: C₄H₁₀O₂S₂
MW: 154.25 g/mol
Kookpunt: ∼125 °C (1013 hPa)
Smeltpunt: 42…44 °C
Vlampunt: >110 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00004877
CAS nummer: 3483-12-3
EINECS: 222-468-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5022472c3f3baef75a703791c548fc6504df810f&vid=81529732db015cb66c1f106092309ae5714325b7&prid=web&clid=SAPDAM