Dithiothreitol (DTT, Cleland's reagent)
Leverancier: Serva
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
cis-2,3-Dihydroxy-1,4-butanedithiol, (R*,R*)-1,4-Dimercapto-2,3-butandiol, DTT, DL-1,4-Dithiothreitol
39759.01
39759.02
39759.03
SERA39759.01EA
29
EUR
SERA39759.01
SERA39759.02
SERA39759.03
Dithiothreitol (DTT, Cleland's reagent)
Dithiothreitol (DTT, Cleland's reagent)
Formule:
C₄H₁₀O₂S₂ MW: 154.25 g/mol Kookpunt: ∼125 °C (1013 hPa) Smeltpunt: 42…44 °C Vlampunt: >110 °C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00004877 CAS nummer: 3483-12-3 EINECS: 222-468-7 |