Glycerine (vegetable based), anhydrous, EMPROVE® EXPERT USP, JP, ACS, Ph. Eur., BP for biopharmaceutical production, SAFC®
Leverancier: MERCK PRODUCTION CHEMICALS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: 1,2,3-Propanetriol, Propane-1,2,3-triol, Glycerol
1.37028.2500 1.37028.9025 1.37028.9950 1.37028.9010 1.37028.1000
1.37028.2500EA 324 EUR
1.37028.2500 1.37028.9025 1.37028.9950 1.37028.9010 1.37028.1000
Glycerine (vegetable based), anhydrous, EMPROVE® EXPERT USP, JP, ACS, Ph. Eur., BP for biopharmaceutical production, SAFC®
Glycerine
Formule: HOCH₂CH(OH)CH₂OH
Kookpunt: 290 °C (1013 hPa)
Dichtheid: 1,26 g/cm³ (20 °C)
Vlampunt: 177 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004722
CAS nummer: 56-81-5
EINECS: 200-289-5
Merck Index: 13,04497
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3668cce85af1464db21fd098526252076b567956&vid=86a15453b54e7106c121d32e4e716385003c4507&prid=web&clid=SAPDAM