1-Butanesulphonyl chloride 98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
403320250 403321000
ACRO403320250EA 113 EUR
ACRO403320250 ACRO403321000
1-Butanesulphonyl chloride 98%
1-Butanesulphonyl chloride
Formule: C₄H₉ClO₂S
Kookpunt: 96 °C (21 mmHg)
Smeltpunt: –25 °C
Dichtheid: 1,2 g/cm³
Vlampunt: 79 °C
MDL Nummer: MFCD00007464
CAS nummer: 2386-60-9
EINECS: 219-204-8
UN: 3265
ADR: 8,II
Order Now
Specification Test Results
Appearance Colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4540 to 1.4570 (20°C, 589 nm)
GC ≥97.5 %