Ashing furnace for crucibles
Leverancier: CEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
306056 307030 307000 306080 306076 306078
CEMA306056EA 0 EUR
CEMA306056 CEMA307030 CEMA307000 CEMA306080 CEMA306076 CEMA306078
Ashing furnace for crucibles
Ovens Verassings- en moffelovens
Microwave ashing system and ashing furnace assemblies. Ash samples with unmatched speed, safety, ease-of use, and energy savings with the Phoenix™ Microwave Muffle Furnace. This system will help improve your process control, allow for rapid adjustments, and reduce out-of-specification products. The temperature ramping capabilities of the Phoenix system mean it can be left idle in a non-heated state. This results in significant energy savings versus conventional muffle furnaces that are kept constantly in a high temperature state.
  • Up to 10 times faster than conventional muffle furnaces
  • Ash up to 15 samples at one time
  • No external hot surfaces prevents risk of injury
  • Uses any type of crucible normally used in conventional ashing systems, even platinum
  • Meets requirements of electrical and microwave muffle furnaces
Place samples into the furnace at room temperature due to the Phoenix’s unique ability to rapidly heat and cool the furnace (protections from rapid sample decomposition and dangerous flames / heat). The units have a closed exhaust air system which minimises contact with toxic or corrosive exhaust gases and integrated safety switches which monitor and prevent overheating.
Bestelinformatie: Different configurations are available, please contact your local Avantor office for details.