NucleoSpin® RNA kit
Leverancier: MACHEREY-NAGEL
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
740955.250
740955.50
740955.10
MANA740955.250EA
1440
EUR
MANA740955.250
MANA740955.50
MANA740955.10
NucleoSpin® RNA kit
Reagentia voor nucleïnezuren
Nucleic Acid Purification Kits and Reagents
NucleoSpin® RNA mini spin kit for the isolation of RNA of the highest integrity.
- rDNase included for on-column DNA digestion– easy and efficient removal of contaminating DNA
- NucleoSpin® filters (shredders) – efficient sample homogenization and reduction of viscosity
- Up to 70 μg ready to use RNA
- Parallel purification of genomic DNA by using the NucleoSpin® RNA/DNA buffer set
Leveringsinformatie: Kit includes lysis buffer RA1, wash buffer RAW2, wash buffer RA3 (concentrate), membrane desalting buffer, reaction buffer for rDNase, rDNase, RNase-free (lyophilised), RNase-free H2O, NucleoSpin® filters (violet rings), NucleoSpin® RNA columns (light blue rings – plus collection tubes), collection tubes (2 ml), collection tubes (1,5 ml) and user manual.