Safety shoes, lace-up, Basic, Jazek
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
12521/36 12521/37 12521/38 12521/39 12521/40 12521/41 12521/42 12521/43 12521/44 12521/45 12521/46 12521/47 12521/48
ELTN12521/36EA 60.9 EUR
ELTN12521/36 ELTN12521/37 ELTN12521/38 ELTN12521/39 ELTN12521/40 ELTN12521/41 ELTN12521/42 ELTN12521/43 ELTN12521/44 ELTN12521/45 ELTN12521/46 ELTN12521/47 ELTN12521/48
Safety shoes, lace-up, Basic, Jazek
Schoenen en laarzen
Durable safety shoes made from cowhide leather and hydrophobised textile Cordura® (tear and abrasion resistant, dirt and water repellent), reflective materials, skin-friendly textile lining, a full padded bellows tongue and collar padding and a dual-density PU/PU NEW BASIC sole (oil, fuel and abrasion resistant, heat resistant up to approx. 130 °C).
  • Climate regulating, breathable textile lining
  • Heel pocket lining made from sturdy abrasion resistant microfibre lining
  • Full-length inlay sole
  • Anti-static soft fleece insole
  • PU toe protection cap
  • Puncture resistant steel midsole
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black/blue
Certificaten: EN ISO 20345; S3, SRC