L(-)-Glutathione (reduced form)
Leverancier: Chemicon
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CHMCAB5520EA
0
EUR
CHMCAB5520
L(-)-Glutathione (reduced form)
L(-)-Glutathione (reduced form)
Formule:
C₁₀H₁₇N₃O₆S MW: 307,33 g/mol Smeltpunt: 185…195 °C Dichtheid: 1,475 g/cm³ (20 °C) Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00065939 CAS nummer: 70-18-8 EINECS: 200-725-4 Merck Index: 13,04488 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...