L(-)-Glutathione (reduced form)
Leverancier: Roth Carl
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: L-Glutathione Reduced, GSH, (S)-2-Amino-5-(((R)-1-((carboxymethyl)amino)-3-mercapto-1-oxopropan-2-yl)amino)-5-oxopentanoic acid, Glutathione Reduced
6382.1 6382.2
ROTH6382.1EA 0 EUR
ROTH6382.1 ROTH6382.2
L(-)-Glutathione (reduced form)
L(-)-Glutathione (reduced form)
Formule: C₁₀H₁₇N₃O₆S
Smeltpunt: 185…195 °C
Dichtheid: 1,475 g/cm³ (20 °C)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00065939
CAS nummer: 70-18-8
EINECS: 200-725-4
Merck Index: 13,04488