L(-)-Glutathione (reduced form), Supelco®
Leverancier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
49739-10ML
49739-50ML
SUPL49739-10MLEA
0
EUR
SUPL49739-10ML
SUPL49739-50ML
L(-)-Glutathione (reduced form), Supelco®
L(-)-Glutathione (reduced form)
Formule:
C₁₀H₁₇N₃O₆S Smeltpunt: 185…195 °C Dichtheid: 1,475 g/cm³ (20 °C) Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00065939 CAS nummer: 70-18-8 EINECS: 200-725-4 Merck Index: 13,04488 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...