Pivalic acid ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

A10776.0C A10776.0E A10776.36
A10776.0CEA 492 EUR
A10776.0C A10776.0E A10776.36
Pivalic acid ≥99%
Pivalic acid

Purity: 99% CAS No.: 75-98-9 Molecular Formula: (CH3)3CCO2H Formula Weight: 102.13 Flash Point: 63°(145°F) Odour: Pungent Boiling Point: 163-165° Solubility: Soluble in alcohol, ether. Slightly soluble in water Beilstein Reference: 969480 Merck Reference: 14,7511 UN No.: 3261 MDL No.: MFCD00004194

Formule: C₅H₁₀O₂
Kookpunt: 163…165 °C
Smeltpunt: 31…35 °C
Dichtheid: 0,91
Vlampunt: 63 °C (145 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004194
CAS nummer: 75-98-9
EINECS: 200-922-5
UN: 3261
ADR: 8,III
Merck Index: 14,7511

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR