4-Methylvaleric acid ≥99%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
A15405.06
A15405.14
A15405.22
A15405.06EA
24.9
EUR
A15405.06
A15405.14
A15405.22
4-Methylvaleric acid ≥99%
4-Methylvaleric acid
Purity: 99%
CAS No.: 646-07-1
Molecular Formula: (CH3)2CH(CH2)2CO2H
Formula Weight: 116.16
Flash Point: 97°(206°F)
Refractive Index: 1.4146
Odour: Unpleasant
Melting Point: -35°
Boiling Point: 199-201°
Beilstein Reference: 1741912
UN No.: 2922
MDL No.: MFCD00002803
Formule:
C₆H₁₂O₂ Kookpunt: 199…201 °C Smeltpunt: –35 °C Dichtheid: 0,923 Vlampunt: 97 °C (206 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00002803 CAS nummer: 646-07-1 EINECS: 211-464-0 ADR: 8,III |