Isopropyl isothiocyanate

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

TL01933.10 TL01933.50
MAYBTL01933.10EA 0 EUR
MAYBTL01933.10 MAYBTL01933.50
Isopropyl isothiocyanate
Isopropyl isothiocyanate
Formule: C₄H₇NS
Kookpunt: 136…137 °C (1013 hPa)
Dichtheid: 0,9475 g/cm³ (20 °C)
MDL Nummer: MFCD00037830
CAS nummer: 2253-73-8
EINECS: 218-851-3
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR