2-Amino-2-methylpropanol, (max. 5% H₂O) 95%
Leverancier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Purity: 95%
CAS No.: 124-68-5
Molecular Formula: C4H11NO
Formula Weight: 89.14
Flash Point: 67°(152°F)
Refractive Index: 1.4455
Odour: Amine-like
pH: 11.3
Melting Point: 24-28°
Boiling Point: 164-166°
Beilstein Reference: 505979
Merck Reference: 14,449
MDL No.: MFCD00008051
Formule:
(CH₃)₂C(NH₂)CH₂OH Kookpunt: 164…166 °C Smeltpunt: 24…28 °C Dichtheid: 0,934 Vlampunt: 67 °C (152 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00008051 CAS nummer: 124-68-5 EINECS: 204-709-8 Merck Index: 14,449 |