tert-Butylmethylether 99+% for HPLC
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
MTBE, 2-Methoxy-2-methylpropane, Methyl-t-Butyl Ether
40477.K2
40477.M1
40477.K2EA
97.1
EUR
40477.K2
40477.M1
tert-Butylmethylether 99+% for HPLC
tert-Butylmethylether
Purity: 99+%
CAS No.: 1634-04-4
Molecular Formula: (CH3)3COCH3
Formula Weight: 88.15
Flash Point: -25°(-13°F)
Physical Form: Liquid
Density: 0.742
Refractive Index: 1.3690
Odour: Mild
Melting Point: -109°
Boiling Point: 55-56°
Beilstein Reference: 1730942
Merck Reference: 14,6032
UN No.: 2350
MDL No.: MFCD00008812
Chromatography solvent
Formule:
(CH₃)₃COCH₃ Kookpunt: 55…56 °C Smeltpunt: –109 °C Dichtheid: 0,742 Vlampunt: –25 °C (–13 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00008812 CAS nummer: 1634-04-4 EINECS: 216-653-1 UN: 2350 ADR: 3,II Merck Index: 14,6032 |