tert-Butylmethylether ≥99.0% (by GC), EMPLURA®, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: MTBE, 2-Methoxy-2-methylpropane, Methyl-t-Butyl Ether
M-Clarity™ quality level = MQ200
1.01843.2500 1.01843.6025 1.01843.6190 1.01843.9011
1.01843.2500EA 93.5 EUR
1.01843.2500 1.01843.6025 1.01843.6190 1.01843.9011
tert-Butylmethylether ≥99.0% (by GC), EMPLURA®, Supelco®
tert-Butylmethylether
Formule: (CH₃)₃COCH₃
MW: 88.15 g/mol
Kookpunt: 55.2
Smeltpunt: –108.6 °C
Dichtheid: 0.7405 g/cm³ (20 °C)
Vlampunt: –28 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008812
CAS nummer: 1634-04-4
EINECS: 216-653-1
UN: 2398
ADR: 3 II
Merck Index: 13,06059
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1712bc714ba0a06e0de37f7cf4e5117256f84425&vid=e1b314f829136075329c50f9e5a593c19ae28b4f&prid=web&clid=SAPDAM