2-Methyl-1-propaanol 99+% for HPLC
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
iso-Butyl alcohol, Isobutylalcohol, iso-Butanol, 2-Methyl-1-propanol
22908.K2
22908.K7
22908.M1
22908.K2EA
73.1
EUR
22908.K2
22908.K7
22908.M1
2-Methyl-1-propaanol 99+% for HPLC
2-Methyl-1-propaanol
Purity: 99+%
CAS No.: 78-83-1
Molecular Formula: C4H10O
Formula Weight: 74.12
Flash Point: 28°(82°F)
Physical Form: Liquid
Density: 0.802
Refractive Index: 1.3960
Appearance: Colourless
Odour: Sweet
pH: 7
Melting Point: -108°
Boiling Point: 107-108°
Beilstein Reference: 1730878
Merck Reference: 14,5131
UN No.: 1212
MDL No.: MFCD00004740
Formule:
(CH₃)₂CHCH₂OH MW: 74.12 g/mol Kookpunt: 107…108 °C Smeltpunt: –108 °C Dichtheid: 0.802 Vlampunt: 28 °C (82 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00004740 CAS nummer: 78-83-1 EINECS: 201-148-0 UN: 1212 ADR: 3 III Merck Index: 14,5131 |