tert-Butylalcohol 99+% ACS
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Trimethylcarbinol, tert-Butyl alcohol, 2-Methyl-2-propanol
33278.AK
33278.K2
33278.K7
33278.M1
33278.AKEA
31
EUR
33278.AK
33278.K2
33278.K7
33278.M1
tert-Butylalcohol 99+% ACS
tert-Butylalcohol
Purity: 99+%
CAS No.: 75-65-0
Molecular Formula: C4H10O
Formula Weight: 74.12
Flash Point: 11°(52°F)
Physical Form: Liquid
Density: 0.786
Refractive Index: 1.3870
Appearance: Colourless
Melting Point: 25°
Boiling Point: 83°
Beilstein Reference: 906698
Merck Reference: 14,1542
UN No.: 1120
MDL No.: MFCD00004464
Formule:
(CH₃)₃COH MW: 74.12 g/mol Kookpunt: 83 °C Smeltpunt: 25 °C Dichtheid: 0.786 Vlampunt: 11 °C (52 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00004464 CAS nummer: 75-65-0 EINECS: 200-889-7 UN: 1120 ADR: 3 II Merck Index: 14,1542 |