Isoamyl alcohol
Leverancier: Serva
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
39557.01
39557.02
SERA39557.01EA
39
EUR
SERA39557.01
SERA39557.02
Isoamyl alcohol
Isoamyl alcohol
Formule:
(CH₃)₂CHCH₂CH₂OH Kookpunt: 130 °C (1013 hPa) Smeltpunt: –117 °C Dichtheid: 0,812 g/cm³ (20 °C) Vlampunt: 42 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00002934 CAS nummer: 123-51-3 EINECS: 204-633-5 UN: 1105 ADR: 3,III Merck Index: 13,05214 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...