3-Methylheptane ≥97%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
L12006.06
L12006.03
L12006.14
L12006.06EA
99.1
EUR
L12006.06
L12006.03
L12006.14
3-Methylheptane ≥97%
3-Methylheptane
Purity: 97%
CAS No.: 589-81-1
Molecular Formula: C8H18
Formula Weight: 114.23
Flash Point: 7°(44°F)
Refractive Index: 1.3980
Melting Point: -121°
Boiling Point: 118-120°
Beilstein Reference: 1730777
UN No.: 1262
MDL No.: MFCD00027244
Formule:
C₈H₁₈ Kookpunt: 118…120 °C Smeltpunt: –121 °C Dichtheid: 0,710 Vlampunt: 7 °C (44 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00027244 CAS nummer: 589-81-1 EINECS: 209-660-6 UN: 1262 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...