1-Naphthylacetic acid 95%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Planofix, 1-Naphthaleneacetic acid
349640250 349641000 349645000
ACRO349640250EA 26.5 EUR
ACRO349640250 ACRO349641000 ACRO349645000
1-Naphthylacetic acid 95%
1-Naphthylacetic acid
Formule: C₁₂H₁₀O₂
MW: 186.21 g/mol
Kookpunt: 352
Smeltpunt: 126…133 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004046
CAS nummer: 86-87-3
EINECS: 201-705-8
Merck Index: 12,06458
Order Now
Specification Test Results
Appearance White to yellow to beige Powder
Infrared spectrum Conforms
Melting point 126 to 133 °C
GC ≥94 %