3-Cyclopentylpropionic acid ≥98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
A18192.14
A18192.22
A18192.14EA
55.9
EUR
A18192.14
A18192.22
3-Cyclopentylpropionic acid ≥98%
3-Cyclopentylpropionic acid
Purity: 98%
CAS No.: 140-77-2
Molecular Formula: C8H14O2
Formula Weight: 142.20
Flash Point: 47°(117°F)
Refractive Index: 1.4570
Boiling Point: 130-133°/12mm
UN No.: 1993
MDL No.: MFCD00001392
Formule:
C₈H₁₄O₂ Kookpunt: 130…133 °C (12 mmHg) Dichtheid: 0,996 Vlampunt: 47 °C (117 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00001392 CAS nummer: 140-77-2 EINECS: 205-433-0 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...