Triphenylacetic acid 99%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
140300050 140300100
ACRO140300050EA 59.7 EUR
ACRO140300050 ACRO140300100
Triphenylacetic acid 99%
Triphenylacetic acid
Formule: C₂₀H₁₆O₂
Kookpunt: 437 °C (1013 hPa)
Smeltpunt: 268…269 °C
Dichtheid: 1,27 g/cm³ (20 °C)
MDL Nummer: MFCD00004185
CAS nummer: 595-91-5
EINECS: 209-873-4
Order Now
Specification Test Results
Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 267 to 275 °C
Titration with NaOH ≥98.5 %