Bisacrylamide, 4x crystallised
Leverancier: BIOMOL RESEARCH LABORATORIES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Bis (N,N'-methylenebisacrylamide), N,N-Methylenebisacrylamide, N,N'-Methylenediacrylamide, N,N’-Methylenebisacrylamide, N,N'-Methylenebisacrylamide, BIS
54538-1 54538-3
BMOL54538-1EA 0 EUR
BMOL54538-1 BMOL54538-3
Bisacrylamide, 4x crystallised
Bisacrylamide
Formule: (H₂C=CHCONH)₂CH₂
Kookpunt: 125 °C (3,33 hPa)
Smeltpunt: 185 °C
Dichtheid: 1,235 g/cm³ (20 °C)
Vlampunt: 138 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00008625
CAS nummer: 110-26-9
EINECS: 203-750-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c1b72b0c3cd5bb916c65f9e2337c5c3b05ea1c91&vid=368f6bf0bc8e0b35ffc7a57d85f5dd210ff0b409&prid=web&clid=SAPDAM