Antifoam agent, Entschäumer S, neodisher®
Leverancier: Dr. Weigert
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
430130 430148
148-3357EA 423 EUR
148-3357 WEIG430148
Antifoam agent, Entschäumer S, neodisher®
Reinigingsmiddelen Detergenten en oppervlaktereinigers
This defoaming agent, liquid concentrate, prevents foaming even at low temperatures a small dose is sufficient. Based on emulsified silicone oils.