Iodoethylene tech. 85%, Technical Grade

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

L12656.03 L12656.06
L12656.03EA 111 EUR
L12656.03 L12656.06
Iodoethylene tech. 85%, Technical Grade
Iodoethylene

Purity: 85% CAS No.: 593-66-8 Molecular Formula: H2C=CHI Formula Weight: 153.95 Refractive Index: 1.5300 Boiling Point: 55-56° MDL No.: MFCD00039404

Formule: C₂H₃I
Kookpunt: 55…56 °C
Dichtheid: 2,080
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00039404
CAS nummer: 593-66-8
EINECS: 209-801-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR