4-Hydroxy-3-methoxycinnamic acid (Ferulic acid)
Leverancier: Roth Carl
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
3-(4-Hydroxy-3-methoxyphenyl)acrylic acid, NSC 2821, NSC 674320, NSC 51986, Coniferic acid, Ferulic acid
9936.1
9936.2
ROTH9936.1EA
0
EUR
ROTH9936.1
ROTH9936.2
4-Hydroxy-3-methoxycinnamic acid (Ferulic acid)
4-Hydroxy-3-methoxycinnamic acid (Ferulic acid)
Formule:
C₁₀H₁₀O₄ MW: 194.18 g/mol |
MDL Nummer:
MFCD00004400 CAS nummer: 1135-24-6 EINECS: 208-679-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...