trans-2,3-Dimethoxycinnamic acid ≥98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
trans-3-(2,3-Dimethoxyphenyl)-2-propenoic acid
A15936.09
A15936.18
A15936.30
A15936.09EA
37
EUR
A15936.09
A15936.18
A15936.30
trans-2,3-Dimethoxycinnamic acid ≥98%
trans-2,3-Dimethoxycinnamic acid
Purity: 98+%
CAS No.: 7345-82-6
Molecular Formula: C11H12O4
Formula Weight: 208.21
Melting Point: 179-183°
Beilstein Reference: 2695122
Formule:
C₁₁H₁₂O₄ |
MDL Nummer:
MFCD00004376 CAS nummer: 7345-82-6 |