N-Bromosuccinimide

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

3261.1 3261.2
ROTH3261.1EA 0 EUR
ROTH3261.1 ROTH3261.2
N-Bromosuccinimide
N-Bromosuccinimide
Formule: C₄H₄BrNO₂
Kookpunt: 339 °C (1013 hPa)
Smeltpunt: 174…179 °C
Dichtheid: 2,098 g/cm³ (20 °C)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00005510
CAS nummer: 128-08-5
EINECS: 204-877-2
UN: 3261
ADR: 8,III
Merck Index: 13,01423

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR