N-Chlorosuccinimide for synthesis, Sigma-Aldrich®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
M-Clarity™ quality level = MQ200
8.02811.0005 8.02811.0250
8.02811.0005EA 32.3 EUR
8.02811.0005 8.02811.0250
N-Chlorosuccinimide for synthesis, Sigma-Aldrich®
N-Chlorosuccinimide
Formule: C₄H₄ClNO₂
Kookpunt: 216,5 °C (1013 hPa)
Smeltpunt: 148…151 °C
Dichtheid: 1,65 g/cm³ (20 °C)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00005511
CAS nummer: 128-09-6
EINECS: 204-878-8
UN: 3261
ADR: 8,II
Merck Index: 13,02183
Order Now
Specification Test Results
Solubility 14 g/l (20 °C)
Melting Point 146 - 151 °C
Molar Mass 133.53 g/mol
Bulk Density 600 kg/m3
Boiling Point 216.5 °C
Flash Point 110 °C
Density 1.65 g/cm3 (20 °C)
pH (20 g/l, H2O) acidic
Hazard Class 8
Hazard Class Description Corrosive material