Pyrrolidine for synthesis, Sigma-Aldrich®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
M-Clarity™ quality level = MQ200
8.07494.0005 8.07494.0250 8.07494.1000
8.07494.0005EA 21.9 EUR
8.07494.0005 8.07494.0250 8.07494.1000
Pyrrolidine for synthesis, Sigma-Aldrich®
Pyrrolidine
Formule: C₄H₉N
Kookpunt: 87 °C (1013 hPa)
Smeltpunt: –63 °C
Dichtheid: 0,86 g/cm³ (20 °C)
Vlampunt: 3 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00005249
CAS nummer: 123-75-1
EINECS: 204-648-7
UN: 1922
ADR: 3 (8),II
Merck Index: 12,08199
Order Now
Specification Test Results
Solubility (20 °C) soluble
Melting Point -63 °C
Molar Mass 71.12 g/mol
Boiling Point 86 - 88 °C (1013 hPa)
Vapor Pressure 65 hPa (20 °C)
Flash Point 3 °C
Refractive Index 1.4435 (20 °C, 589 nm)
Explosion Limit 1.6 - 10.6 %(V)
Density 0.86 g/cm3 (20 °C)
pH 12.9 (100 g/l, H2O, 20 °C)
Ignition Point 345 °C
Saturation concentration 466 g/m3 (20 °C) Air
Hazard Class 3
Hazard Class Description Flammable Liquid