N-Methyl-2-pyrrolidon (NMP), watervrij ≥99.5%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Packaged under argon in resealable ChemSeal™ bottles.
43741.AE
43741.K2
43741.M1
43741.5Y
43741.AEEA
76.9
EUR
43741.AE
43741.K2
43741.M1
43741.5Y
N-Methyl-2-pyrrolidon (NMP), watervrij ≥99.5%
N-Methyl-2-pyrrolidon (NMP)
Purity: 99.5%
CAS No.: 872-50-4
Molecular Formula: C5H9NO
Formula Weight: 99.13
Flash Point: 86°(187°F)
Physical Form: Liquid
Density: 1.033
Refractive Index: 1.4705
Sensitivity: Light Sensitive and Hygroscopic
Appearance: Colourless to pale yellow
Odour: Amine-like
Melting Point: -24°
Boiling Point: 204°
Beilstein Reference: 106420
Merck Reference: 14,6117
MDL No.: MFCD00003193
Formule:
C₅H₉NO Kookpunt: 204 °C Smeltpunt: –24 °C Dichtheid: 1,033 Vlampunt: 86 °C (187 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00003193 CAS nummer: 872-50-4 EINECS: 212-828-1 Merck Index: 14,6117 |