N-Methyl-2-pyrrolidon (NMP) 99,0 ACS
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
1-Methyl-2-pyrrolidinone, N-Methyl-2-pyrrolidinone,NMP, 1-Methyl-2-pyrrolidone, NMP, Methyl Pyrrolidone
43894.K2
43894.AP
43894.M1
43894.K2EA
110
EUR
43894.K2
43894.AP
43894.M1
N-Methyl-2-pyrrolidon (NMP) 99,0 ACS
N-Methyl-2-pyrrolidon (NMP)
Purity: 99.0+%
CAS No.: 872-50-4
Molecular Formula: C5H9NO
Formula Weight: 99.13
Flash Point: 86°(187°F)
Physical Form: Liquid
Density: 1.033
Refractive Index: 1.4705
Sensitivity: Light Sensitive and Hygroscopic
Appearance: Colourless to pale yellow
Odour: Amine-like
Melting Point: -24°
Boiling Point: 204°
Beilstein Reference: 106420
Merck Reference: 14,6117
MDL No.: MFCD00003193
Formule:
C₅H₉NO MW: 99.13 g/mol Kookpunt: 204 °C Smeltpunt: –24 °C Dichtheid: 1.033 Vlampunt: 86 °C (187 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00003193 CAS nummer: 872-50-4 EINECS: 212-828-1 Merck Index: 14,6117 |